missing translation for 'onlineSavingsMsg'
Learn More

ITPA Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA588421

Product Code. 16526671

  • 472.00€ / 100µL
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Immunogen sequence: MAASLVGKKI VFVTGNAKKL EEVVQILGDK FPCTLVAQKI DLPEYQGEPD EISIQKCQEA VRQVQGPVLV EDTCLCFNAL GGLPGPYIKW FLEKLKPEGL HQLLAGFEDK SAYALCTFAL STGDPSQPVR LFRGRTSGRI VAPRGCQDFG WDPCFQPDGY EQTYAEMPKA EKNAVSHRFR ALLELQEYFG SLAA; Positive Samples: U-87MG, HT-29, Mouse small intestine, Mouse heart, Rat liver; Cellular Location: Cytoplasm

Pyrophosphatase that hydrolyzes the non-canonical purine nucleotides inosine triphosphate (ITP), deoxyinosine triphosphate (dITP) as well as 2'-deoxy-N-6-hydroxylaminopurine triposphate (dHAPTP) and xanthosine 5'-triphosphate (XTP) to their respective monophosphate derivatives. The enzyme does not distinguish between the deoxy- and ribose forms. Probably excludes non-canonical purines from RNA and DNA precursor pools, thus preventing their incorporation into RNA and DNA and avoiding chromosomal lesions.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ITPA
Polyclonal
Unconjugated
ITPA
2010016I08Rik, AU020102, C20orf37, dJ794I6.3, HAMAP-Rule:MF_03148}, HLC14-06-P, inosine triphosphatase, inosine triphosphatase (nucleoside triphosphate pyrophosphatase), inosine triphosphatase {ECO:0000255, inosine triphosphate pyrophosphatase, inosine triphosphate pyrophosphatase {ECO:0000255, inosine triphosphate pyrophosphohydrolase, Itp, ITPA, ITPase, ITPase {ECO:0000255, My049, My049 protein, Non-canonical purine NTP pyrophosphatase, non-canonical purine NTP pyrophosphatase {ECO:0000255, non-standard purine NTP pyrophosphatase, non-standard purine NTP pyrophosphatase {ECO:0000255, NTPase, NTPase {ECO:0000255, nucleoside triphosphate pyrophosphatase, nucleoside-triphosphate diphosphatase, nucleoside-triphosphate diphosphatase {ECO:0000255, Nucleoside-triphosphate pyrophosphatase, nucleoside-triphosphate pyrophosphatase {ECO:0000255, OK/SW-cl.9, Putative oncogene protein hlc14-06-p
Rabbit
Affinity Chromatography
RUO
16434, 311422, 3704
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.58 mg/mL
PBS with 50% glycerol and 0.01% thimerosal, pH 7.3
D3ZW55, Q9BY32, Q9D892
ITPA
Recombinant fusion protein containing a sequence corresponding to amino acids 1-194 of human ITPA (NP_2584121)
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers