Learn More
Jenway™ pH Electrodes
Use these electrodes for advanced applications where faster response times and greater reproducibility are required.
Brand: Jenway™ 68006-37
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Quickly reach equilibrium
- Significantly reduced response times
- Far greater reproducibility
- Significantly reduced long-term drift
Especificaciones
2mm plug | |
Refillable Reference with Bulb End | |
in lab with colloidal suspensions, samples containing iodides, samples that are high in solids, viscous solutions and samples containing proteins | |
Glass |
12 mm | |
12 dia. x 120 mm L | |
120 mm | |
0°C to 50°C |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Jenway™ pH Electrodes > Electrode type: refillable reference with bulb end; Connector: 2mm plug; For in lab with colloidal suspensions, samples containing iodides, samples that are high in solids, viscous solutions and samples containing proteins
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido