Learn More
3M™ Jupiter™ Intrinsically Safe (IS) Pouches
3M™ Jupiter™ Intrinsically Safe (IS) Pouches are a replacement part for the 3M™ Jupiter™ Powered Air Respirator. They must be used in combination with an IS battery pack. Suitable for operations where flammable or combustible materials present a risk of fire or explosion.
Brand: 3M™ 7000000138
78.16 EUR valid until 2025-06-21
Use promo code "24097" to get your promotional price.
Description
- Product Type: Replacement Pouch for Intrinsically Safe (IS) PAPR System
- Description: Suitable for use in potentially flammable or explosive atmospheres
- Material (Coating): Explosive-Protective Coating
- For Use With (Equipment): 3M™ Jupiter™ Powered Air Respirator (Turbo)
- For Use With (Application): Respiratory Protection
Specifications
Suitable for use in potentially flammable or explosive atmospheres | |
Respiratory Protection | |
Explosive-protective Coating |
Replacement Pouch | |
3M Jupiter Powered Air Respirator (Turbo) | |
1 Ea. |
Promotions
-
Multiple Manufacturers
Outlet Corner
Use code 24097 to receive this promotion.Promo Code: 24097Expires: 2025-06-16
Days
15Hours
16Minutes
17Seconds
18