Learn More
uvex K1 Ear Muffs
The Uvex K protective ear muffs offer an impressive combination of high levels of protection combined with great design.
Brand: uvex 2600001
23.27 EUR valid until 2025-03-09
Use promo code "24130" to get your promotional price.
This item is not returnable.
View return policy
Description
- Soft surface
- Easy to mould
- Comfortable to wear
- Low weight
Specifications
EN 352-1 | |
No | |
Reusable | |
Black | |
Over-the-head |
29 dB | |
Ear Muffs | |
Yes | |
No | |
No |
Product Content Correction
Product Title
uvex K1 Ear Muffs > Color: Black, Green