Learn More
Kaolin, pure, Thermo Scientific Chemicals Product Code.: 10772441
Kaolin, pure, Thermo Scientific Chemicals
Product Code. 10772441
1 kg, Plastic Bottle
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
Product Code. 10772441
Brand: Thermo Scientific Acros 211740010
Request Bulk or Custom Format
Edge
CAS: 1332-58-7 | Al2H4O9Si2 | 258.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1332-58-7
Molecular Formula Al2H4O9Si2
Molecular Weight (g/mol) 258.16
MDL Number MFCD00062311
InChI Key NLYAJNPCOHFWQQ-UHFFFAOYSA-N
Synonym kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss
PubChem CID 56841936
IUPAC Name oxo-oxoalumanyloxy-[oxo(oxoalumanyloxy)silyl]oxysilane;dihydrate
SMILES O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O
Specifications
Melting Point 1750.0°C
Color Beige
Physical Form Powder
Quantity 1 kg
Merck Index 14,5282
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in cold acids and alkali hydroxides
Formula Weight 258.2
Grade Pure
Loss on Drying 1.5% max.
Loss on Ignition 10.8% to 11.8%
Packaging Plastic Bottle
Chemical Name or Material Kaolin
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465235 Certificate TypeCertificate of Analysis Date19/08/2024 Catalog Number211740025,211740100,211740010
Lot NumberA0459927 Certificate TypeCertificate of Analysis Date08/02/2024 Catalog Number211740025,211740100,211740010
Lot NumberA0456188 Certificate TypeCertificate of Analysis Date15/09/2023 Catalog Number211740025,211740100,211740010
Lot NumberA0454625 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number211740025,211740100,211740010
Lot NumberA0449731 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog Number211740025,211740100,211740010
Product Title
Kaolin, pure, Thermo Scientific Chemicals >