missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
229.00€ - 477.00€
Specifications
Antigen | KCNB1 |
---|---|
Dilution | Western Blot 5 - 10 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18302446
|
Bio-Techne
NBP3-11900-25UG |
25 μg |
229.00€
25µg |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
18392583
|
Bio-Techne
NBP3-11900-100UG |
100 μg |
477.00€
100µg |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
Description
KCNB1 Polyclonal antibody specifically detects KCNB1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
KCNB1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Delayed rectifier potassium channel 1, delayed rectifier potassium channel Kv2.1, DRK1, h-DRK1, h-DRK1 K(+) channel, Kv2.1, potassium channel protein DRK1, potassium voltage-gated channel subfamily B member 1, potassium voltage-gated channel, Shab-related subfamily, member 1, Voltage-gated potassium channel subunit Kv2.1 | |
Rabbits were immunized with a synthetic peptide derived from the rat Kv2.1 potassium channel conjugated to KLH (Uniprot: Q14721) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 5 - 10 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS | |
3745 | |
IgG | |
Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title