Learn More
Thermo Scientific™ KingFisher™ Plastics for 24 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Brand:  Thermo Scientific™ 97002620B
581.40 EUR valid until 2025-06-30
Use promo code "24108" to get your promotional price.
 View more versions of this product
Product Code. 17100079
  • 698.00€ / Pack of 50
Description
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24 DW tip comb, plate, sterile
KingFisher Apex
50/Cs.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2420210 Certificate TypeCertificate of Compliance Date10/10/2024 Catalog Number97002620B
Lot Number2222510 Certificate TypeCertificate of Compliance Date19/08/2022 Catalog Number97002620B
Lot Number2221910 Certificate TypeCertificate of Compliance Date19/08/2022 Catalog Number97002620B
Lot Number2221510 Certificate TypeCertificate of Compliance Date19/08/2022 Catalog Number97002620B
Lot Number2222210 Certificate TypeCertificate of Compliance Date19/08/2022 Catalog Number97002620B
Special Offers
Special Offers
  • Invitrogen , ABI Applied Biosystems , Thermo Scientific , Affymetrix eBioscience
    Savings to support your research.
    Promo Code: 24108
    Expires2025-06-27
    Days
    37
    Hours
    2
    Minutes
    55
    Seconds
    57
    New Offer
    Save Now
Product Content Correction
Product Title
Thermo Scientific™ KingFisher™ Plastics for 24 deep-well format >