missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VACUUBRAND™ KF DN 10mm Stainless Steel Small Flange With Female Ground Joint
56.55€ - 65.20€
Specifications
Material | Stainless Steel |
---|---|
Quantity | 1 Each |
Size (Metric) | KF DN 10 mm |
Type | Small Flange |
Description
Specifications
Stainless Steel | |
1 Each | |
KF DN 10 mm | |
Small Flange |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title