Learn More
Novus Biologicals™ Kidney Matched Tumor and Normal Tissue Lysate
Two matched 1 mg/mL vials (100 μg/vial) of human kidney tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
Brand:  Novus Biologicals™ NBP2-47103-2VIAL
Product Code. 18158168
  • 655.00€ / Pack of 2
Estimated Shipment: 19-03-2025
Description
The Kidney Matched Tumor and Normal Tissue Lysate is derived from human.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 Vials
Store at -80C. Avoid freeze-thaw cycles.
Documents
One moment while we fetch your results.
Product Title
Novus Biologicals™ Kidney Matched Tumor and Normal Tissue Lysate > 2 Vial