missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ KIMAX™ Automatic Burets with Bottles
Class A
Brand: DWK Life Sciences 17124F-10
This item is not returnable.
View return policy
Description
standard taper joints fit into reagent bottles with standard taper necks.
- Constructed with overflow tubes that return directly to reservoir
- For air intake, overflow cups have hose connection for attaching drying tube; second drying tube provided for use between rubber bulb and reservoir
- Precision bore tubing permanently marked with individual serial number
- Durable black ceramic enamel scale
- Inset shows top configuration
Certifications
A certificate is provided to indicate instrument traceability to standards provided by NIST™.

Specifications
10 mL | |
Borosilicate Glass | |
Volumetric Analysis | |
Reservoir Bottle, U-Shaped Drying Tube, Vented Connecting Tube, Rubber Squeeze Bulb, No. 1 Single-Holed Rubber Stopper, PTFE Stopcock Plug and 1/4 In. I.D. Rubber Tubing, Durable Black Ceramic Enamel Scale | |
29 | |
±0.02 mL | |
Automatic Buret | |
ASTM E438 |
765 mm | |
0.05 mL | |
30.1 in. | |
Black | |
PTFE | |
1000 mL | |
Class A |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction