Learn More
Kimberly-Clark Professional™ Toilet Tissue Dispenser
Robust and lockable, offering a refillable, reliable anytime top up solution. Single sheet dispensing promotes a hygienic washroom and reduces cross contamination.
Brand: Kimberly-Clark Professional™ 8972
112.22 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Designed to work in harmony with the trusted and familiar Kleenex™ and Scott™ brands.
- These washroom dispensers are designed to support hygiene standards and deliver an exceptional washroom solution suited to your workplace hygiene needs.
- Our Kimberly-Clark toilet tissue dispenser, with its sleek and contemporary design, is the ideal toilet tissue dispenser to create a luxurious and modern washroom image.
- This single sheet toilet tissue dispenser and its effective single sheet dispensing ensures you only touch the product you use, helping to promote high standards of washroom hygiene and reduce cross contamination between users.
- This stainless steel toilet tissue dispenser provides a refillable, reliable, anytime top up solution for washroom maintenance.
- Refilling the dispenser is simple and the Hassle-free maintenance keeps costs down. The robust toilet paper dispenser also features a hidden lock for refilling.
- This commercial toilet tissue dispenser measures 31.9 cm (L) x 17.5 cm (W) x 14 cm (D)
- The Kimberly-Clark toilet paper dispenser is compatible with a variety of Kleenex™ and Scott™ folded toilet tissue packs, recommended: 8408, 8042. It is also compatible with: 4471, 4477, 8407, 8577.
Specifications
Toilet Paper Dispenser | |
Toilet Tissue | |
Wall Mount | |
Hinged Lid with Front Opening |
Silver | |
Stainless Steel | |
1 |
Product Content Correction
Product Title
Kimberly-Clark Professional™ Toilet Tissue Dispenser > Material: Stainless Steel