Learn More
DWK Life Sciences Kimble™ Ultra-Ware™ Fritted Glass Support Base
The Kimble™ Ultra-Ware™ Fritted Glass Support Base is supplied with a 40/35 joint and glass frit. It is suitable for use with the Kimble™ Ultra-Ware™ 90 mm Microfiltration Assembly.
Brand: DWK Life Sciences 953826-0090
This item is not returnable.
View return policy
Description
- Product Type: Fritted Support Base
- Material: Glass
- Joint Size: 40/35
- For Use with: Kimble™ Ultra-Ware™ 90 mm Microfiltration Assembly 40/35
Specifications
Not Autoclavable | |
Glass Frit | |
Glass | |
40/35 |
Reusable | |
Kimble Ultra-Ware 90 mm Microfiltration Assembly 40/35 | |
Non-sterile | |
Fritted Support Base |
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Ultra-Ware™ Fritted Glass Support Base > Fritted Glass Support Base