Learn More
DWK Life Sciences Kimble™ Kontes™ Pellet Pestle™
Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Brand:  DWK Life Sciences 749516-0500
90.45 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 11888502
  • 209.00€ / Each
Only null left
Estimated Shipment: 01-09-2025
Description
Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Disposable Pellet Pestle
  • Choose autoclavable polypropylene or RNase-, DNase-, pyrogen-free material (manufactured under M5.5 [Class 10,000] cleanroom conditions)
  • Available with or without 100 microtubes
  • Microtubes have the same properties and come individually wrapped to ensure purity
  • Length of pestle with tip, 7cm (2.75 in.)
Reusable Pellet Pestle
  • Autoclavable CTFE polymeric head with a stainless-steel shaft
  • Pestle length with tip, 6cm (2.5 in.) for 0.5mL size; 14cm (5.59 in.) for 1.5mL size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
0.5 mL
Pellet Pestle
Resuspend Protein and DNA pellets or grind soft tissue in microcentrifuge tubes
No
500ÎĽL and 1.5mL microcentrifuge tube
6mm
0.01 oz.
CTFE/Stainless Steel
Reusable
0.23 in.
w/o Microtubes
6mm
0.01 oz., 0.5mL
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Kontes™ Pellet Pestle™ > Reusable Pellet Pestle without Microtubes; CTFE/Stainless Steel; 0.5mL