Learn More
DWK Life Sciences™ Kimble™ Score-Break Trimmed-Stem Ampules
Offer maximum solution stability, easy sealing and opening
80.30€
Product Code | Brand | Capacity (Metric) | Color | Diameter (English) Outer | Length (English) Body | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Color | Diameter (English) Outer | Length (English) Body | Price | Quantity & Availability | |||
17760858
|
DWK Life Sciences™
12040G-2 |
2 mL | Amber | 0.47 in. | 1.18 in. |
80.30€
Pack of 144 |
Estimated Shipment: 15-12-2025
Log in to see stock. |
|||
Description
- Choice of Amber 203 or Kimax-51™ clear
- United States Pharmacopeia (USP) Type I borosilicate glass
- Pull sealed
- Score-break, trimmed stem for easy opening
- Special ampule file not required
Amber ampules
- 1mL and 2mL
Kimax-51™ ampules
- 2mL and 20mL
Specifications
Borosilicate Glass | |
Prescored Ampule |
Product Title
DWK Life Sciences™ Kimble™ Score-Break Trimmed-Stem Ampules