Learn More
HMC Europe Vertical Autoclave Baskets
With this item you can easily and safely bring your sterilized goods into and out of the autoclave; we offer baskets in various sizes, with and without bowl or closed cans with and without holes.
Brand:  HMC Europe 51009
 View more versions of this product
Product Code. 10309253
  • 312.00€ / Each
Estimated Shipment: 08-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with the HMC-HG and HMC-HV autoclaves.
Specifications
Specifications
345 mm
305 mm
33.8 cm
80 L (Total), 50 L (Nominal)
9 in.
Wire mesh basket
338 x 235 mm
Wire mesh basket
13.3 in.
13.2/21.13 gal.
HG 50/80 autoclave
235 mm
Stainless Steel