missing translation for 'onlineSavingsMsg'
Learn More
L-m-Tyrosine, 97+%, Thermo Scientific Chemicals Product Code.: 11362219

L-m-Tyrosine, 97+%, Thermo Scientific Chemicals

Product Code. 11362219
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11362219

Brand: Thermo Scientific Alfa Aesar H27224.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An unnatural amino acid, that has demonstrated potential in treating Parkinsons disease, Alzheimers disease, and arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide, dilute acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerated (4°C).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587-33-7
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00063059
InChI Key JZKXXXDKRQWDET-SVGMAFHSNA-N
Synonym l-m-tyrosine, 3-hydroxy-l-phenylalanine, meta-tyrosine, s-2-amino-3-3-hydroxyphenyl propanoic acid, 3-tyrosine, 3-hydroxyphenylalanine, m-tyrosine, l-m-tyr, unii-d5yf57v4qw, 3-m-hydroxyphenyl alanineShow MoreShow Less
PubChem CID 6950578
ChEBI CHEBI:44303
IUPAC Name (2S)-2-amino-3-(3-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=CC(O)=C1)C(O)=O

Specifications

Melting Point 260°C to 270°C
Solubility Information Soluble in dimethyl sulfoxide,dilute acid.
Quantity 1 g
Formula Weight 181.19
Percent Purity ≥97%
Chemical Name or Material L-m-Tyrosine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252539 Certificate TypeCertificate of Analysis Date08/11/2024 Catalog NumberH27224.03,H27224.MD
Lot Number10190743 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27224.03,H27224.MD
Lot Number10167852 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27224.03,H27224.MD
Lot Number10228871 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27224.03,H27224.MD
Lot Number10218386 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27224.03,H27224.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-m-Tyrosine, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.