missing translation for 'onlineSavingsMsg'
Learn More

L-Quisqualic acid, Tocris Bioscience™

Product Code. 30000707
Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

AMPA agonist; also group I mGlu agonist

L-Quisqualic acid is a glutamate receptor agonist acting at AMPA receptors and metabotropic glutamate receptors positively linked to phosphoinositide hydrolysis. L-Quisqualic acid sensitizes neurons in hippocampus to depolarization by L-AP6 (the so called 'quis' effect).

Chemical Identifiers

CAS 52809-07-1
Molecular Formula C5H7N3O5
Molecular Weight (g/mol) 189.13
MDL Number MFCD00069337
InChI Key ASNFTDCKZKHJSW-REOHCLBHSA-N
Synonym quisqualic acid, quisqualate, l-quisqualic acid, +-quisqualic acid, 3h quisqualate, unii-8oc22c1b99, 2s-2-amino-3-3,5-dioxo-1,2,4-oxadiazolidin-2-yl propanoic acid, 2jbk, 1,2,4-oxadiazolidine-2-propanoic acid, alpha-amino-3,5-dioxo-, s, 3-3,5-dioxo-1,2,4-oxadiazolidin-2-yl-l-alanine
PubChem CID 40539
ChEBI CHEBI:8734
IUPAC Name (2S)-2-amino-3-(3,5-dioxo-1,2,4-oxadiazolidin-2-yl)propanoic acid
SMILES N[C@@H](CN1OC(=O)NC1=O)C(O)=O

Specifications

Quantity 10 mg
Formula Weight 189.13
Percent Purity >99%
Chemical Name or Material L-Quisqualic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.