Learn More
Thermo Scientific Chemicals L-Tyrosine, 99%
32.70€ - 285.00€
Chemical Identifiers
CAS | 60-18-4 |
---|---|
Molecular Formula | C9H11NO3 |
Molecular Weight (g/mol) | 181.19 |
MDL Number | MFCD00002606 |
InChI Key | OUYCCCASQSFEME-SVGMAFHSNA-N |
Synonym | l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l |
PubChem CID | 6057 |
ChEBI | CHEBI:17895 |
SMILES | N[C@@H](CC1=CC=C(O)C=C1)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11419053
|
Thermo Scientific Chemicals
A11141.18 |
50 g |
32.70€
50g |
In Stock
Log in to see stock. |
|||||
11438840
|
Thermo Scientific Chemicals
A11141.22 |
100 g |
55.60€
100g |
In Stock
Log in to see stock. |
|||||
11429053
|
Thermo Scientific Chemicals
A11141.30 |
250 g |
91.30€
250g |
In Stock
Log in to see stock. |
|||||
11409053
|
Thermo Scientific Chemicals
A11141.0B |
1000 g |
285.00€
1000g |
In Stock
Log in to see stock. |
|||||
Description
L-Tyrosine is used in pharmaceuticals, dietary supplements and food additives. It is a precursor to alkaloids such as morphine, neurotransmitters, epinephrine, p-coumaric acid, thyroxine, pigment melanin and catcholamines. It plays a vital role in photosynthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Tyrosine is used in pharmaceuticals, dietary supplements and food additives. It is a precursor to alkaloids such as morphine, neurotransmitters, epinephrine, p-coumaric acid, thyroxine, pigment melanin and catcholamines. It plays a vital role in photosynthesis.
Solubility
Soluble in hydrochloric acid. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and strong reducing agents.
Chemical Identifiers
60-18-4 | |
181.19 | |
OUYCCCASQSFEME-SVGMAFHSNA-N | |
6057 | |
N[C@@H](CC1=CC=C(O)C=C1)C(O)=O |
C9H11NO3 | |
MFCD00002606 | |
l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l | |
CHEBI:17895 |
Specifications
>300°C | |
99% | |
MFCD00002606 | |
14,9839 | |
Soluble in hydrochloric acid. Slightly soluble in water. | |
N[C@@H](CC1=CC=C(O)C=C1)C(O)=O | |
181.19 | |
CHEBI:17895 | |
99% | |
L-Tyrosine |
60-18-4 | |
C9H11NO3 | |
392441 | |
l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l | |
OUYCCCASQSFEME-SVGMAFHSNA-N | |
(2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid | |
6057 | |
181.19 | |
1.46 g/mL |
Safety and Handling
GHS H Statement
H315-H319-H335
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 200-460-4
missing translation for 'rtecsNumber' : YP2275600
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only