Learn More
Epredia™ Lab Vision™ NY-BR-1 Mouse Monoclonal Antibody
Take advantage of a quality product that delivers accurate results with the Epredia™ NY-BR-1, Mouse Monoclonal Antibody.
270.00€ - 1690.00€
Specifications
Antigen | NY-BR-1 |
---|---|
Clone | NY-BR-1 No.2 |
Applications | Immunohistochemistry |
Conjugate | Unconjugated |
Host Species | Mouse |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | |||||
12610487
|
Epredia™
MS-1932-P0 |
Purified | 100 ÎĽL | N/A | ||||||
12600487
|
Epredia™
MS-1932-P |
Purified | 1 mL |
1690.00€
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12620487
|
Epredia™
MS-1932-R7 |
Ready-to-Use | 7 mL | N/A | ||||||
12640487
|
Epredia™
MS-1932-RQ |
Ready-to-Use | 12 mL | N/A | ||||||
Description
NY-BR-1 is a novel differentiation marker predominantly expressed in normal breast and breast cancer. It has been shown by Immunohistochemistry (IHC) that NY-BR-1 was present solely in ductal epithelium of normal breast tissue. Invasive carcinoma of the breast and carcinoma in situ were positive for NY-BR-1, whereas most other tumors and normal tissues are negative, with the exceptions of one-third of sweat gland carcinomas and around two percent of prostate carcinoma. NY-BR-1 is reported to express more frequently in estrogen-positive and lymph node-negative primary carcinomas, and more commonly in grade 1 than in grade 2 or grade 3. In a study by Varga et al (2006) forty-nine percent of lymph node metastasis were shown to be positive.Host Species: Mouse
Clone: NY-BR-1No. 2
Isotype: IgG1/Îş
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: a 77 aa peptide from human NYBR-1 protein
Molecular Weight: 150-160kDa
Positive Control: Breast carcinoma
Cellular Localization: Predominantly cytoplasmic and occasional nuclear
Recommended for:
Immunohistochemistry (Formalin/paraffin)
Specifications
NY-BR-1 | |
Immunohistochemistry | |
Mouse | |
Primary |
NY-BR-1 No.2 | |
Unconjugated | |
a 77 aa peptide from human NYBR-1 protein |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.