missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Placental Alk. Phosphatase (PLAP) Ab-4, Mouse Monoclonal Antibody

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ Placental Alk. Phosphatase (PLAP) Ab-4, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-734-S

 View more versions of this product

Product Code. 12632197

  • 505.00€ / 1mL
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Host Species: Mouse

Clone: 8A9

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human Placental Alkaline Phosphatase (PLAP)

Molecular Weight: 60-70kDa

Positive Control: Placenta or seminoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Placental Alk. Phosphatase (PLAP) Ab-4
Monoclonal
Unconjugated
Purified human Placental Alkaline Phosphatase (PLAP)
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
8A9
Mouse
1 mL
Cancer and Tumor Biology
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.