Learn More
Labconco™ Protector XVS™ Blower Transition Adapter for Ventilation Stations
Blower Transition Adapter for Ventilation Stations
Brand:  Labconco™ 4861700
Product Code. 15479454
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • For use with Labconco Protector XVS Ventilation Stations
  • Powder-coated steel blower transition adapter allows round PVC duct to be connected to the exhaust side of the blower
Specifications
Specifications
4.25 x 7.75 in. round duct, Protector™ VS ventilation station
1.4 Kg
Epoxy-coated Steel
3 lb.
Blower Transition Adapter
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Labconco™ Protector XVS™ Blower Transition Adapter for Ventilation Stations > Blower Transition Adapter