Learn More
Kirsch Philipp™ Laboratory Freezer
Re-circulating air cooling, a rapid defrosting system, quick-freeze function.
Brand: Kirsch Philipp™ 10050
This item is not returnable.
View return policy
Description
- Re-circulating air cooling
- Optimized ventilation plates to minimize interior temperature differences
- Rapid freezing cycle
- LED temperature display
- Automatical defrosting and melt water evaporation
- Easy accessible control panel with key switch
- Micro-processor controlled temperature and thawing regulation to maximize temperature stability
- Minimum/maximum temperature memory: optical and acoustical alarm
- RS-485 interface for computer based temperature documentation
- Potential free contact for alarm transmission
- Lockable door
Specifications
Laboratory Freezer | |
-5°C to -20°C | |
24 to 40 kg | |
57 x 60 x 140 cm | |
50/60 Hz | |
170 kg (gross) |
500 L | |
5 Adjustable | |
Storage of laboratory chemicals | |
7.6kWh/24 hr. | |
640 w |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Kirsch Philipp™ Laboratory Freezer > Laboratory Freezer