missing translation for 'onlineSavingsMsg'
Learn More

Elma™ Laboratory Glass Holder Set

Elmasonic EASY accessory

Brand:  Elma™ 1049786

Product Code. 18018083

  • 71.75€ / Each
Estimated Shipment: 06-06-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For holding laboratory bottles with a neck diameter of up to 40 mm
  • Stainless steel tripod rod with a length of 600 mm
Specificatie

Specificatie

Tube Holder
Elmasonic EASY
Product Suggestions

Product Suggestions

Video's
SDS
Documentatie

Documentatie

Certificates
Speciale aanbiedingen

Speciale aanbiedingen

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Elma™ Laboratory Glass Holder Set > For use with Elmasonic EASY

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.