man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Facility Safety and Maintenance
Signs and Tags
Facilities Maintenance Signs
Brady™ Laminated Polyester Prohibition Signs: Eating and Drinking Prohibited
Print
Brady™ Laminated Polyester Prohibition Signs: Eating and Drinking Prohibited
3.35€ - 4.70€
Specifications
Shape
Round
View More Specs
Products
2
Certificates
Products
2
Product Code
Brand
Diameter (Metric)
Price
Quantity & Availability
Product Code
Brand
Diameter (Metric)
Price
Quantity & Availability
10171974
missing translation for 'viewDocuments'
missing translation for 'viewProductCertificates'
Brady™
250049
100 mm
3.35€
Each
In Stock
Log in
to see stock.
Add to basket
10181974
missing translation for 'viewDocuments'
missing translation for 'viewProductCertificates'
Brady™
250050
200 mm
4.70€
Each
In Stock
Log in
Specifications
Shape
Round
Certificates
Back to Top