Learn More
Thermo Scientific Chemicals Laminin (925-933)
For cellular and molecular biology applications
Brand: Thermo Scientific Chemicals J64340.|D
Description
Derived from residues 925-933 of human laminin B1, this peptide binds the laminin receptor, Inhibits experimental metastasis. find useful for product applications. And it is also used in Western Blot, Blocking Peptides, Dot Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Flow Cytomety, Cell Culture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
Solid | |
967.05 | |
0.5 mg | |
Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg |
-30°C to -10°C | |
Derived from residues 925-933 of human laminin B1, this peptide binds the laminin receptor | |
Laminin (925-933) | |
RUO - research use only |
RUO – Research Use Only