Learn More
Thermo Scientific Chemicals Laminin A Chain (2091-2108)
For cellular and molecular biology applications
106.53€ - 169.85€
Specifications
Purification Method | HPLC |
---|---|
Content And Storage | -30°C to -10°C |
Format | Solid |
Molecular Weight (g/mol) | 2016.30 |
Product Type | Laminin A Chain (2091-2108) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15457009
|
Thermo Scientific Chemicals
J65640.!D |
0.5 mg |
106.53€
0.50mg |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
15467009
|
Thermo Scientific Chemicals
J65640.M^ |
1 mg |
169.85€
1mg |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
Description
Supports neurite outgrowth and stimulates neuronal-like process formation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
Solid | |
Laminin A Chain (2091-2108) | |
Cys-Ser-Arg-Ala-Arg-Lys-Gln-Ala-Ala-Ser-Ile-Lys-Val-Ala-Val-Ser-Ala-Asp-Arg |
-30°C to -10°C | |
2016.30 | |
RUO - research use only |
RUO – Research Use Only