Learn More
Carl Zeiss™ Replacement Bulb for Microscope
Use these bulbs with Carl Zeiss microscopes
Brand: Carl Zeiss™ 000000-0482-730
This item is not returnable.
View return policy
Description
Average lifetime over 2,000 hrs.Specifications
Mercury Short-arc HXP 120 (D) Bulb | |
Average lifetime 2,000 hours | |
Zeiss Microscopes | |
1 Ea. |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Carl Zeiss™ Replacement Bulb for Microscope > Type: HXP 120