Learn More
OHAUS™ Large Three Prong Sleeves
OHAUS™ Large Three Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.
Brand:  OHAUS™ 30400261
7.76 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 15702679
  • 10.25€ / Each
In Stock
Description
  • Available in red or black vinyl and fiberglass
  • Fiberglass sleeves are ideal for application above 100°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3
Vinyl
Red
Prong Sleeve
Large
Documents
One moment while we fetch your results.
Product Title
OHAUS™ Large Three Prong Sleeves > Marerial: Vinyl; Color: Red