Learn More
Kartell™ Latex Gas Sampling Balloon
Gass collection bags that are constructed from latex. Kartell™ Latex Gas Sampling Balloon are best suited for collecting, containing, and carrying gas samples.
Brand: Kartell™ 0123300
31.45 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
Latex balloons for gas samplingSpecifications
4 L | |
Latex | |
4000 cc | |
200 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
23Hours
12Minutes
54Seconds
57
Product Content Correction
Product Title
Kartell™ Latex Gas Sampling Balloon > 200mm dia.