Learn More
Kartell™ Latex Gas Sampling Balloon
Gass collection bags that are constructed from latex. Kartell™ Latex Gas Sampling Balloon are best suited for collecting, containing, and carrying gas samples.
Brand:  Kartell™ 0123100
21.13 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 10456123
  • 22.25€ / Each
Estimated Shipment: 30-05-2025
Description
Latex balloons for gas sampling
Specifications
Specifications
1.5 L
Latex
1500 cc
150 mm
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    7
    Hours
    11
    Minutes
    44
    Seconds
    42
Product Content Correction
Product Title
Kartell™ Latex Gas Sampling Balloon > 150mm dia.