Learn More
Kartell™ Latex Gas Sampling Balloon
Gass collection bags that are constructed from latex. Kartell™ Latex Gas Sampling Balloon are best suited for collecting, containing, and carrying gas samples.
Brand:  Kartell™ 0123000
12.04 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
 View more versions of this product
Product Code. 10680374
  • 16.75€ / Each
Estimated Shipment: 17-06-2025
Description
  • Cost effective
  • Ideal for collecting, containing, and carrying gas samples
Specifications
Specifications
Gas Sampling Balloon
1 L
Latex
130 mm
Special Offers
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Kartell™ Latex Gas Sampling Balloon > Capacity, Metric: 1L