missing translation for 'onlineSavingsMsg'
Learn More

LCN6 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

198.00€ - 468.00€

Specifications

Antigen LCN6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18392427
View Documents
Bio-Techne
NBP3-04897-20UL
20 μL
198.00€
20µL
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
18396246
View Documents
Bio-Techne
NBP3-04897-100UL
100 μL
468.00€
100µL
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
Description

Description

LCN6 Polyclonal specifically detects LCN6 in Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

LCN6
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
158062
IgG
Affinity purified
Western Blot
Unconjugated
Rabbit
Mouse, Rat
epididymal-specific lipocalin LCN6, epididymal-specific lipocalin-6, hLcn5, LCN5, lipocalin 5, lipocalin 6, lipocalin-5, UNQ643
Recombinant fusion protein containing a sequence corresponding to amino acids 25-120 of human LCN6 (NP_945184.1).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.