missing translation for 'onlineSavingsMsg'
Learn More

Linalyl acetate, 95%, synthetic, Thermo Scientific Chemicals

20.40€ - 68.94€

Chemical Identifiers

CAS 115-95-7
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.28
MDL Number MFCD00008907
InChI Key UWKAYLJWKGQEPM-UHFFFAOYSA-N
Synonym linalyl acetate, linalool acetate, bergamiol, bergamol, linalol acetate, lynalyl acetate, licareol acetate, bergamot mint oil, acetic acid linalool ester, 3,7-dimethyl-1,6-octadien-3-ol acetate
PubChem CID 8294
ChEBI CHEBI:78333
IUPAC Name 3,7-dimethylocta-1,6-dien-3-yl acetate
SMILES CC(=CCCC(C)(C=C)OC(=O)C)C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10378080
View Documents Promotion Details
Thermo Scientific Acros
232300050
5 mL Glass bottle
20.40€
5mL
In Stock
to see stock.
Add to basket
 
10490121
View Documents
Thermo Scientific Acros
232301000
100 mL Glass bottle
25.13€
100mL
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
 
10400131
View Documents
Thermo Scientific Acros
232305000
500 mL Glass bottle
68.94€
500mL
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

115-95-7
196.28
UWKAYLJWKGQEPM-UHFFFAOYSA-N
8294
3,7-dimethylocta-1,6-dien-3-yl acetate
C12H20O2
MFCD00008907
linalyl acetate, linalool acetate, bergamiol, bergamol, linalol acetate, lynalyl acetate, licareol acetate, bergamot mint oil, acetic acid linalool ester, 3,7-dimethyl-1,6-octadien-3-ol acetate
CHEBI:78333
CC(=CCCC(C)(C=C)OC(=O)C)C

Specifications

115-95-7
0.9010g/mL
85°C
94% min. (GC)
(CH3)2=CHCH2CH2C(CH3)(OOCCH3)CH=CH2
MFCD00008907
0.901
Solubility in water: insoluble in water. Other solubilities: soluble in ethanol,alcohol,ether,diethyl phthal,mineral oil,fixed oils
CC(=CCCC(C)(C=C)OC(=O)C)C
196.28
CHEBI:78333
95%
Linalyl acetate, 95%
Colorless
220°C
Authentic
C12H20O2
1.4490 to 1.452
15, 5551
linalyl acetate, linalool acetate, bergamiol, bergamol, linalol acetate, lynalyl acetate, licareol acetate, bergamot mint oil, acetic acid linalool ester, 3,7-dimethyl-1,6-octadien-3-ol acetate
UWKAYLJWKGQEPM-UHFFFAOYSA-N
3,7-dimethylocta-1,6-dien-3-yl acetate
8294
196.28
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 204-116-4

missing translation for 'rtecsNumber' : RG5910000

missing translation for 'tsca' : TSCA

Product Identifier
  • Linalyl acetate, synthetic

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.

Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only