Learn More
Novus Biologicals™ LIPN Overexpression Lysate
Brand:  Novus Biologicals™ NBP2-08240
Product Code. 18057307
  • 700.00€ / 0.10mg
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The LIPN Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_195939. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens LIPN, mRNA.
Native