missing translation for 'onlineSavingsMsg'
Learn More

Lithium aluminum oxide, Thermo Scientific Chemicals

101.51€ - 20290.95€

Chemical Identifiers

CAS 12003-67-7
Molecular Formula AlLiO2
Molecular Weight (g/mol) 65.92
MDL Number MFCD00016172
InChI Key YQNQTEBHHUSESQ-UHFFFAOYSA-N
Synonym lithium aluminate, lithium metaaluminate, aluminum lithium dioxide, aluminate alo21-, lithium, lithium aluminum spinal, lithium aluminate lialo2, hsa 10, aluminum lithium oxide, aluminum lithium oxide lialo2, lithium aluminum oxide lialo2
PubChem CID 123268
IUPAC Name lithium;oxido(oxo)alumane
SMILES [Li+].[O-][Al]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11386738
View Documents
Thermo Scientific Alfa Aesar
013412.18
50 g
101.51€
50g
In Stock
to see stock.
Add to basket
 
11396738
View Documents
Thermo Scientific Alfa Aesar
013412.30
250 g
503.51€
250g
In Stock
to see stock.
Add to basket
 
11306748
View Documents
Thermo Scientific Alfa Aesar
013412.A1
1 kg
20290.95€
1kg
In Stock
to see stock.
Add to basket
 
Description

Description

It is used as an inert electrolyte support material in molten carbonate fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12003-67-7
65.92
YQNQTEBHHUSESQ-UHFFFAOYSA-N
123268
[Li+].[O-][Al]=O
AlLiO2
MFCD00016172
lithium aluminate, lithium metaaluminate, aluminum lithium dioxide, aluminate alo21-, lithium, lithium aluminum spinal, lithium aluminate lialo2, hsa 10, aluminum lithium oxide, aluminum lithium oxide lialo2, lithium aluminum oxide lialo2
lithium;oxido(oxo)alumane

Specifications

>1625°C
White
AlLiO2
lithium aluminate, lithium metaaluminate, aluminum lithium dioxide, aluminate alo21-, lithium, lithium aluminum spinal, lithium aluminate lialo2, hsa 10, aluminum lithium oxide, aluminum lithium oxide lialo2, lithium aluminum oxide lialo2
YQNQTEBHHUSESQ-UHFFFAOYSA-N
lithium;oxido(oxo)alumane
123268
Odorless
Lithium aluminum oxide
12003-67-7
Powder
MFCD00016172
Insoluble in water.
[Li+].[O-][Al]=O
65.92
65.92
2.62
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-434-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.