Learn More
Copan Italia™ Loop in Resealable Pouch
Use Copan Italia™ Loop in Resealable Pouch as a flexible inoculator for brittle agar. The light blue loop is disposable, calibrated and flexible.
Brand: Copan Italia™ 179CSR40
This item is not returnable.
View return policy
Description
- DIsposable
- Calibrated
- 4000Pack
- Light Blue
Specifications
Single-use | |
Flexible | |
Light Blue | |
4000/Pk. |
1 μL | |
Gamma-irradiated | |
Inoculating Loop | |
Yes |