Learn More
Kartell™ Polypropylene Low Form Beakers with Printed Blue Graduations
Click to view available options
Capacity: 500mL
Description
Manufactured from highly translucent polypropylene for durability, clarity and good chemical resistance. Autoclavable Kartell™ Polypropylene Low Form Beakers with Printed Blue Graduations provide an economical and safe solution for stirring, mixing and dispensing liquids in the laboratory.
- Permanent printed blue graduations provide clarity and reproducibility
- Autoclavable at 121°C for 10 minutes
- Withstands 100°C of continuous work
- Class B, Low form
- Complies with ISO 7056-1981(E) and BS 5404 Part 1
- Suitable for foodstuff
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T24S00632 | Declaration of Conformity | 01/08/2024 | 0182500 |
T23S02376 | Declaration of Conformity | 28/06/2024 | 0182500 |
T22S01505 | Declaration of Conformity | 05/12/2023 | 0182500 |
T23S02376 | Certificate of Compliance | 03/02/2024 | 0182500 |
T22S01505 | Certificate of Compliance | 26/10/2023 | 0182500 |
Product Content Correction
Product Title
Kartell™ Polypropylene Low Form Beakers with Printed Blue Graduations >