Learn More
LRCH1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-44281G
Product Code. 18720893
  • 639.00€ / 0.10mL
Description
LRCH1 Polyclonal antibody specifically detects LRCH1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LRCH1
Polyclonal
Western Blot
Purified
Western Blot
DyLight 488
The epitope recognized by this antibody maps to a region between residue 325 to 375 of human Leucine-rich repeat and calponin homology domain-containing protein 1 using the numbering given in entry Q9Y2L9.3 (GeneID 23143).
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
LRCH1 Antibody, DyLight 488, Novus Biologicals™ > 0.1 mL; DyLight 488