missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ Luer Lock Instrument Syringe with Adjustable Stop
Ideal for dispensing both liquids and gases.
Brand: Hamilton™ 80223
This item is not returnable.
View return policy
Description
- Precision-machined PTFE plunger which creates a leak-free seal
- Tip essentially wipes the interior of the syringe barrel free of sample
- Reduces the chance that a deposit will occur and cause the plunger to freeze
Specifications
Autoclavable | |
Gas sterilizable | |
1000 psig (68.9 bar) | |
10°C | |
Luer Lock |
Borosilicate Glass, PTFE | |
Gastight Syringe | |
115°C | |
25 μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction