missing translation for 'onlineSavingsMsg'
Learn More

LY 379268, Tocris Bioscience™

Product Code. 30001330
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly selective group II mGlu agonist

LY 379268 is a highly selective group II mGlu receptor agonist (EC50 values are 2.69 and 4.48 nM for hmGlu2 and hmGlu3 respectively) that displays > 80-fold selectivity over group I and group III receptors. Provides protection against NMDA-mediated cell death in vitro and offers almost complete protection against CA1 hippocampal damage following global ischemia in gerbils. Orally and systemically active. LY379268 disodium salt also available.

Chemical Identifiers

CAS 191471-52-0
Molecular Formula C7H9NO5
Molecular Weight (g/mol) 187.151
InChI Key YASVRZWVUGJELU-MDASVERJSA-N
Synonym 1r,4r,5s,6r-4-amino-2-oxabicyclo 3.1.0 hexane-4,6-dicarboxylic acid, d07grg, 4-amino-2-oxa-bicyclo 3.1.0 hexane-4,6-dicarboxylic acid, 1r,4r,5s,6r-4-amino-2-oxa-bicyclo 3.1.0 hexane-4,6-dicarboxylic acid, 1s,2r,5r,6r-2-amino-4-oxabicyclo 3.1.0 hexane-2,6-dicarboxylic acid, 2-oxabicyclo 3.1.0 hexane-4,6-dicarboxylic acid, 4-amino-, 1r,4r,5s,6r-rel
PubChem CID 10197984
IUPAC Name (1R,4R,5S,6R)-4-amino-2-oxabicyclo[3.1.0]hexane-4,6-dicarboxylic acid
SMILES C1C(C2C(C2O1)C(=O)O)(C(=O)O)N

Specifications

Quantity 10 mg
Formula Weight 187.15
Percent Purity >99%
Chemical Name or Material LY 379268
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.