missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lysis Buffer 1 + RNase for Food ID

Recommended for use with the PrepSEQ Nucleic Acid Extraction Kit (4480466) to prepare DNA for meat ID and GMO testing from food and feed samples.

Brand:  Thermo Scientific™ A24401

Product Code. 15666019

  • 376.00€ / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Recommended for use with the PrepSEQ Nucleic Acid Extraction Kit (4480466) to prepare DNA for meat ID and GMO testing from food and feed samples
  • Protocol using Lysis Buffer 1 + RNase in conjunction with the PrepSEQ Nucleic Acid Extraction Kit can be readily automated on the Thermo Scientific™ KingFisher™ mL Food Protection Purification System platform
  • Reduces hands-on time and increases reproducibility of the DNA extraction procedure
  • The purified DNA can then be used in PCR or other molecular biology techniques

Purified DNA is well-suited for use with the following food and feed testing kits:

  • TaqMan GMO Screening Kit (4466334)
  • TaqMan Roundup Ready Soya Quantification Kit (4466335)
  • TaqMan GMO Maize Quantification Kit (4481972)
  • RapidFinder Beef ID Kit (A24391)
  • RapidFinder Pork ID Kit (A24392)
  • RapidFinder Equine ID Kit (A15570)
  • RapidFinder Poultry ID Kit (A24397)
  • RapidFinder Chicken ID Kit (A24393)
  • RapidFinder Turkey ID Kit (A24394)
  • RapidFinder Ruminent ID Kit (A24396)

Includes

  • 4 x 0.5 L bottles of Lysis Buffer 1
  • 2 x 10 mg tubes each of RNase, dried

Store contents at room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

4 x 0.5 L
Food and Environmental Samples
Contains:
• 4 bottles of Lysis Buffer 1
• 2 tubes each of RNase, dried

Store at 15–30°C. After reconstitution with Nuclease-free water, store RNase between -25°C and -15°C.
High-throughput Compatible
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers