Learn More
Sartorius™ MA100Q-000230V1 Moisture Analyzer
Brand:  Sartorius™ MA100Q-000230V1
11410.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11338973
  • 13560.00€ / Each
Estimated Shipment: 12-06-2025
Specifications
Digital
230 V
Product Title
Sartorius™ MA100Q-000230V1 Moisture Analyzer > Dimensions: 156 H x 350 W x 453mm D