Learn More
Emma Safety Footwear Mack M Safety Shoes
A superb shoe for galvanizing and welding metal. A closed front flap protects against stray sparks and hot metal splinters and offers enhanced protection for the metatarsal. Includes polyurethane outer toe protection and a concealed steel toe cap.
Brand: Emma Safety Footwear MM53486341
This item is not returnable.
View return policy
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Duo Heat Resistant Outsole
- Heat and Cold insulated
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
41 |
Laces | |
Mack M | |
Leather | |
Steel |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Emma Safety Footwear Mack M Safety Shoes > Size: 41