Learn More
Magnesium powder, -100+200 mesh, 99.6% (metals basis excluding Ca), Thermo Scientific Chemicals Product Code.: 11393228
Magnesium powder, -100+200 mesh, 99.6% (metals basis excluding Ca), Thermo Scientific Chemicals
Product Code. 11393228
250 g, 250g
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11393228
Brand: Thermo Scientific Alfa Aesar 036194.30
Request Bulk or Custom Format
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
For vacuum deposition
For vacuum deposition
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]
Specifications
Color White
UN Number UN1418
Quantity 250 g
Percent Purity 99.6%
Mesh Size −100+200 Mesh
Odor Odorless
Form Powder
Assay (metals basis excluding Ca), Ca typically 0.1-1%
Chemical Name or Material Magnesium powder
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number62300522 Certificate TypeCertificate of Analysis Date23/05/2025 Catalog Number036194.18,036194.30
Lot Number62500068 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog Number036194.18,036194.30
Lot NumberN14J008 Certificate TypeCertificate of Analysis Date14/07/2023 Catalog Number036194.18,036194.30
Lot NumberT05I005 Certificate TypeCertificate of Analysis Date19/08/2022 Catalog Number036194.18,036194.30
Lot NumberE12U006 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number036194.18,036194.30
Product Title
Magnesium powder, -100+200 mesh, 99.6% (metals basis excluding Ca), Thermo Scientific Chemicals >