Learn More
Rogo Sampaic™ Malassez Counting Cells
For determining the number of particles per volume unit of a liquid. Rogo Sampaic™ Malassez Counting Cells have a simple or double grid with a depth of 0.2mm.
128.00€ - 160.00€
Specifications
Product Type Malassez Cell
Quantity 1Each
Sample Type Blood
Depth (Metric) 0.2mm
View More Specs
Products 2
Product Code Brand Grid Type Price Quantity & Availability  
Product Code Brand Grid Type Price Quantity & Availability  
11740835
View Documents
Rogo Sampaic™
ZMB024
Simple
128.00€
Each
Only null left
Estimated Shipment: 01-07-2025
Add to basket
11720845
View Documents
Rogo Sampaic™
ZMB021
Double
160.00€
Each
Only null left
In Stock
Edge
Add to basket
Description
Description
  • For nematodes or cells in cerebro-spinal fluid
  • Provides for accurate manual measurements
Specifications
Specifications
Malassez Cell
1Each
Blood
0.2mm
Documents
Product Certifications
Product Content Correction
Product Title
Rogo Sampaic™ Malassez Counting Cells