Learn More
Thermo Scientific™ Male Luer-LOK Priming Syringes
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
Brand: Thermo Scientific™ 365KL531
93.39 EUR valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
- Square tip to prevent damage to the injector
- Wide range of volumes
- Precision made from borosilicate glass and stainless steel
- Robust design and easy-to-read markings
Specifications
Gas Tight GC Autosampler Syringe, Luer-Lok | |
1 mL | |
Gas Chromatography Systems | |
365RNL22 |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
52Hours
7Minutes
0Seconds
56 -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
8Hours
7Minutes
0Seconds
56