Learn More
Emma Safety Footwear Mammoet M1966 D Safety Shoes
State-of-the art Sympatex™ construction provides perfect foot protection for 'hands on' roles within the Chemical Industry and Agriculture. Virtually waterproof, these boots allow feet to breathe properly, keeping them cool and dry.
195.00€
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Color | Black |
Design | Mammoet M1966 D |
Liner Material | Hydro-Tec™ Sanitized Silver / Sympatex™ |
Material | Oiled Nubuck |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15958801
|
Emma Safety Footwear
584848-37 |
37 | N/A | ||||||
15968801
|
Emma Safety Footwear
584848-38 |
38 | N/A | ||||||
15978801
|
Emma Safety Footwear
584848-39 |
39 | N/A | ||||||
15988801
|
Emma Safety Footwear
584848-40 |
40 | N/A | ||||||
15998801
|
Emma Safety Footwear
584848-41 |
41 | N/A | ||||||
15908811
|
Emma Safety Footwear
584848-42 |
42 | N/A | ||||||
15918811
|
Emma Safety Footwear
584848-43 |
43 | N/A | ||||||
15928811
|
Emma Safety Footwear
584848-44 |
44 | N/A | ||||||
15938811
|
Emma Safety Footwear
584848-45 |
45 | N/A | ||||||
15948811
|
Emma Safety Footwear
584848-46 |
46 | N/A | ||||||
15958811
|
Emma Safety Footwear
584848-47 |
47 | N/A | ||||||
15968811
|
Emma Safety Footwear
584848-48 |
48 | N/A | ||||||
15978811
|
Emma Safety Footwear
584848-49 |
49 | N/A | ||||||
Description
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Duo Heat Resistant Outsole
- Sympatex™ Waterproof
- Heat and cold insulated
- Sanitized
Specifications
EN ISO 20345:2011 | |
Mammoet M1966 D | |
Oiled Nubuck |
Black | |
Hydro-Tec™ Sanitized Silver / Sympatex™ | |
Steel |