Learn More
Manganese Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical Product Code.: 10518430
Manganese Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical
Product Code. 10518430 Shop All Fisher Chemical Products
100 mL, HDPE plastic bottle
Click to view available options
Quantity:
100 mL
Packaging:
HDPE plastic bottle
Product Code. 10518430
Brand: Fisher Chemical J/8290/05
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Color Colorless
Physical Form Liquid
Quantity 100 mL
MDL Number 11111
Packaging HDPE plastic bottle
compliance-icons
Product Identifier
  • Manganese solution 10 000 ppm in 1M HNO6
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction
Product Title
Manganese Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical >