Learn More
Fisherbrand™ Manual Crimper, Steel, for Aluminum/Steel caps
Description
- Crimper
- Manual
- Steel
- Non-autoclavable
- Adjustable
Specifications
Pneumatic Basic Crimping Tool | |
11 mm | |
Steel | |
Not Autoclavable |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Focus On Chromatography
File size: 75.3 MB
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Product Title
Fisherbrand™ Manual Crimper, Steel, for Aluminum/Steel caps > For 11mm caps