Learn More
Marimastat, Tocris Bioscience™ Product Code.: 30002120
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
Marimastat, Tocris Bioscience™
Product Code. 30002120
10 mg
Click to view available options
Quantity:
1 mg
10 mg
Product Code. 30002120
Brand: Tocris Bioscience 2631/10
Estimated Shipment: 05-03-2025
Broad spectrum MMP inhibitor
Broad spectrum inhibitor of MMPs (IC50 values are 3, 5, 6, 9 and 13 nM for MMP-9, MMP-1, MMP-2, MMP-14 and MMP-7 respectively). Inhibits peritoneal dissemination of human gastric cancer cells in vivo through inhibition of tumor angiogenesis.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 154039-60-8
Molecular Formula C15H29N3O5
Molecular Weight (g/mol) 331.413
InChI Key OCSMOTCMPXTDND-OUAUKWLOSA-N
Synonym marimastat, marimastat usan, marimastat usan/inn, unii-d5eqv23tds, d5eqv23tds, 2s,3r-3-1s-2,2-dimethyl-1-methylcarbamoxy propyl carboyl-2-hydroxy-5-methylhexanohydroxamic acid, 2s,3r-3-1s-2,2-dimethyl-1-methylcarbamoyl propyl carbamoyl-2-hydroxy-5-methylhexanohydroxamic acid, 2s,3r-n 4-2s-3,3-dimethyl-1-methylamino-1-oxobutan-2-yl-n 1 ,2-dihydroxy-3-2-methylpropyl butanediamide, 2r,3s-n-2s-3,3-dimethyl-1-methylamino-1-oxobutan-2-yl-n',3-dihydroxy-2-2-methylpropyl butanediamide, 2s,3r-n-1s-2,2-dimethyl-1-methylcarbamoyl propyl-n',2-dihydroxy-3-2-methylpropyl butanediamideShow MoreShow Less
PubChem CID 119031
ChEBI CHEBI:50662
IUPAC Name (2R,3S)-N-[(2S)-3,3-dimethyl-1-(methylamino)-1-oxobutan-2-yl]-N',3-dihydroxy-2-(2-methylpropyl)butanediamide
SMILES CC(C)CC(C(C(=O)NO)O)C(=O)NC(C(=O)NC)C(C)(C)C
Specifications
Quantity 10 mg
Formula Weight 331.41
Percent Purity >99%
Chemical Name or Material Marimastat
Product Content Correction
Product Title
Marimastat, Tocris Bioscience™ >